missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies STEMdiff™ Astrocyte Differentiation Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Description
- STEMdiff™ Astrocyte Differentiation Kit is used to rapidly and efficiently generate astrocytic precursors from neural progenitor cells (NPCs) derived from human pluripotent stem cells (hPSCs) using STEMdiff™ SMADi Neural Induction Kit (Catalog #08581)
- These astrocytic precursors are then matured further into astrocytes using STEMdiff™ Astrocyte Maturation Kit (Catalog #100-0016)
- Using this system, a highly pure population of astrocytes (> 70% S100B-positive and > 60% GFAP-positive astrocytes; < 15% doublecortin-positive neurons) can be generated from hPSCs in as few as 7 weeks and can be maintained long term in culture
- Cells derived using these products are versatile tools for modeling human neurological development and disease, drug screening, toxicity testing, and cell therapy validation
Specifications
Specifications
| Cell Type | Neural Cells, PSC-Derived, Neural Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture, Differentiation |
| Quantity | 1 Kit |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction