missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Stella Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585971
This item is not returnable.
View return policy
Description
Stella Polyclonal Antibody for Western Blot, IHC (P)
DPPa3 (stella-related protein) is a stem cell marker protein which is restricted to pluripotent human embryonic stem cells, premeiotic germ lineage cells and testicular germ cell tumor cells. Otherwise known as 'developmental pluripotency-associated protein 3,' it is expressed in primordial germ cells, Oocytes, preimplantation embryos and pluripotent cells. This protein has a SAP-like domain and a splicing factor motif-like structure, suggesting possible roles in chromosomal organization or RNA processing.
Specifications
| Stella | |
| Polyclonal | |
| Unconjugated | |
| DPPA3 | |
| 2410075G02Rik; Cap1p; Compaction-associated protein 1; Crg1; developmental pluripotency associated 3; developmental pluripotency-associated 3; developmental pluripotency-associated protein 3; Dppa3; PCG7; Pgc7; primordial germ cell 7; primordial germ cell protein 7; Stella; STELLAR; Stella-related protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 359787, 73708 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| Q6W0C5, Q8QZY3 | |
| DPPA3 | |
| Synthetic peptide within mouse Stella aa 101-150. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction