missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Steel Wire Bag Stand

Product Code. 10384401 Shop All Brand Products
Change view
Click to view available options
Color:
White
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10384401 White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10384401 Supplier Brand™ Supplier No. 759701

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Manufactured from epoxy-coated steel wire. BRAND™ Steel Wire Bag Stand includes rubber feet to prevent slippage. Available with polypropylene or polyamide bags.

TRUSTED_SUSTAINABILITY

Specifications

Description Epoxy coated
Autoclavable Not Autoclavable
Color White
For Use With (Equipment) Disposal bags
Includes 50 polyamide disposal bags
Material Steel Wire
Height (Metric) 250 mm
Diameter (Metric) 12 cm
Product Type Stand
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.