missing translation for 'onlineSavingsMsg'
Learn More

Retsch™ Stainless Steel Collecting Pan for Test Sieves

Collects resulting samples from test sieves. These stainless steel pans are compatible with multiple test sieves and sieve shakers, based upon the required diameter.

Brand:  Retsch™ 200S/STREC

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10585072

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Stainless steel
  • Compatible with multiple test sieves, based upon diameter
Specifications

Specifications

1 Each
Collecting Pan
Test sieve
200mm
Stainless Steel
50mm
Videos