Learn More
Manutan Steel roller bearing conveyor, 90 degree bend, 70kg, 300mm wide
Product Code. 12811499
Encompass_Preferred
Product Code. 12811499 Supplier Manutan Supplier No. A341385
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Conveyor tracks manufactured from high quality 2mm thick steel and supplied complete with zinc plated steel or high impact PVC roller bearings.
  • Roller bearing pitch: 32 Roller bearings
  • Track width: 300mm