Learn More
Manutan Steel framed track support stand, width 450mm, T foot height 650/1100
Product Code. 12882618
Encompass_Preferred
Product Code. 12882618
Brand: Manutan A341427
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Supports should be fitted every 1. 5m and at frame joints. Available as 'T' foot for bolting to floor, rubber foot, or castors with additional knee brace. Adjustable in height (add 100mm height for castors).
  • Track width: for 450mm track width
  • Height: 650mm-1100mm