missing translation for 'onlineSavingsMsg'
Learn More

Stearyl stearate, 98%

Product Code. 10678162 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10678162 5 g Glass bottle 5g
10712653 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10678162 Supplier Thermo Scientific Supplier No. 368110050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2778-96-3 | C36H72O2 | 536.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2778-96-3
Molecular Formula C36H72O2
Molecular Weight (g/mol) 536.97
MDL Number MFCD00056225
InChI Key NKBWPOSQERPBFI-UHFFFAOYSA-N
Synonym stearyl stearate, octadecyl stearate, octadecanoic acid, octadecyl ester, stearic acid stearyl ester, unii-5wx2egd0dk, stearic acid, stearyl ester, octadecanyl octadecanoate, 5wx2egd0dk, stearic acid, octadecyl ester, cyclochem ss
PubChem CID 17720
IUPAC Name octadecyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCCCC

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)16COO(CH2)17CH3
Quantity 5 g
Formula Weight 536.97
Percent Purity 98%
Physical Form Powder or Platelets
Chemical Name or Material Stearyl stearate, 98%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.