missing translation for 'onlineSavingsMsg'
Learn More

Stearoylcarnitine, MedChemExpress

Product Code. 16474758
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16474758 10 mg 10mg
16407497 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16474758 Supplier Medchem Express Supplier No. HY113202
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Stearoylcarnitine, a fatty ester lipid molecule, is a human endogenous metabolite. Stearoylcarnitine acts as a metabolomics biomarker for early-onset-preeclampsia and late-onset-preeclampsia.

Stearoylcarnitine, a fatty ester lipid molecule, is a human endogenous metabolite. Stearoylcarnitine acts as a metabolomics biomarker for early-onset-preeclampsia and late-onset-preeclampsia.

Specifications

Chemical Name or Material Stearoylcarnitine
CAS 25597-09-5
Color White
Purity Grade Notes Research
Molecular Formula C25H49NO4
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (116.92 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 1 mg/mL (2.34 mM; ultrasonic and warming and heat to 80°C)
SMILES CCCCCCCCCCCCCCCCCC(O[C@H](CC([O-])=O)C[N+](C)(C)C)=O
Molecular Weight (g/mol) 427.66
Formula Weight 427.66
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.