missing translation for 'onlineSavingsMsg'
Learn More

Stearic anhydride, 98%

Product Code. 10279460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10279460 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10279460 Supplier Thermo Scientific Acros Supplier No. 419710250
Outlet Corner
Use promo code 28080
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 638-08-4 | C36H70O3 | 550.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 638-08-4
Melting Point 69°C to 71°C
Color Colorless
Density 0.8400g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C36H70O3
Linear Formula (CH3(CH2)16CO)2O
MDL Number MFCD00009004
Quantity 25 g
Specific Gravity 0.84
Synonym stearic anhydride, octadecanoic acid, anhydride, stearic acid anhydride, stearicanhydride, octadecanoic acid, 1,1'-anhydride, octadecanoic anhydride, e r degrees eeaou, octadecanoic anhydride #, acmc-1b4gn, octadecanoic acid,1,1'-anhydride
InChI Key WVJVHUWVQNLPCR-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCCCCCC
IUPAC Name octadecanoyl octadecanoate
Molecular Weight (g/mol) 550.95
PubChem CID 69485
Formula Weight 550.95
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Stearic anhydride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.