missing translation for 'onlineSavingsMsg'
Learn More

Stearic Acid, TRC

Product Code. 17957724
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17957724 10 g 10g
17909354 1 g 1g
17949554 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17957724 Supplier Toronto Research Chemicals Supplier No. TRCS68649510G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-11-4
Molecular Formula C18 H36 O2
Molecular Weight (g/mol) 284.48
Synonym Stearic acid (8CI), 1-Heptadecanecarboxylic acid, 1-Octadecanoic acid, 17FA, 400JB9103-88, 50S, 60R, 60R Rubber Grade Stearic Acid, A 1760, Adeka Fatty Acid SA 910, Adeka SA 300, B 1841, Barolub FTA, Beads Stearic Acid YR, Century 1210, Century 1220, Century 1224, Century 1230, Century 1240, Daiwax STF, Dervacid 3155, Edenor C 18, Edenor C 18/98, Edenor C 18/98-100, Edenor HT-JG 60, Edenor ST 1, Edenor ST 20, Emersol 120, Emersol 153NF, Emersol 6349, Emersol 7036, F 1500, F 3, F 3 (lubricant), FA 1655, G 20, G 270, HST, Humko Industrene R, Hydrofol Acid 150, Hydrofol Acid 1895, Hystrene 5016, Hystrene 80, Hystrene 9718, Hystrene 9718NF, Hystrene 9718NFFG, Hystrene S 97, Hystrene T 70, Imex Stearic Acid ST, Industrene 5016, Industrene 5016K, Industrene 8718, Industrene 9018, Industrene R, Kam 1000, Kam 2000, Kam 3000, Kiri stearic acid, Kortacid 1895, Kortacid 1989, Loxiol G 20, Lunac 30, Lunac S 20, Lunac S 25, Lunac S 30, Lunac S 35, Lunac S 40, Lunac S 50, Lunac S 50V, Lunac S 90, Lunac S 90KC, Lunac S 90V, Lunac S 98, Lunac YA, MXST 100, NAA 173, NAA 173K, NAA 175, NAA 175S, NAA 180, NAA 180P, NAA 180P1, NSC 25956, NSC 261168, Neo-Fat 18, Neo-Fat 18-53, Neo-Fat 18-54, Neo-Fat 18-59, Neo-Fat 18S, Nissan Unister NAA 180, Nopcocera LU 6418, Norsorex AP, PD 185, PF 1808, Palmac 98-18, Parteck LUB-STA 50, Prifac 2918, Prifac 2979, Prifac 2981, Prifrac 2981, Pristerene 4900, Pristerene 4963, Pristerene 9429, Pristerene 9559, Radiacid 0152, Radiacid 0427, Radiacid 444, S 300, S 300 (fatty acid), S 30C, S 30C (fatty acid), S 40T, S 90V, SA 1801, SA 1837, SA 200, SA 400, SA 400 (fatty acid), Selosol 920, Speziol, Stearex Beads, Stearic Acid 50S, Stearic Acid Camellia, Stearic Acid Cherry, Stearic acid 1801, Stearic acid 1840, Stearophanic acid, Sunfat 18S, T 18, T 32, TST, Tsubaki, Unister NAA 180, V 1890, VEG-FGK, VLZ 200, Vanicol, Vis-Plus, WO 2, WO 2 (fatty acid), n-Octadecanoic acid
IUPAC Name octadecanoic acid
SMILES CCCCCCCCCCCCCCCCCC(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)
Formula Weight 284.2715
Chemical Name or Material Stearic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.