missing translation for 'onlineSavingsMsg'
Learn More

Stearic acid, MedChemExpress

Product Code. 16442389
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16442389 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16442389 Supplier Medchem Express Supplier No. HYB2219
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Stearic acid is a long chain dietary saturated fatty acid which exists in many animal and vegetable fats and oils.

Stearic acid is a long chain dietary saturated fatty acid which exists in many animal and vegetable fats and oils.

Specifications

Chemical Name or Material Stearic acid
CAS 57-11-4
Color White
Purity Grade Notes Research
Molecular Formula C18H36O2
Quantity 1 g
Solubility Information DMSO : 125 mg/mL (439.40 mM; Need ultrasonic)
SMILES CCCCCCCCCCCCCCCCCC(O)=O
Molecular Weight (g/mol) 284.48
Formula Weight 284.48
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.