Learn More
Stearic acid, 98%, Thermo Scientific Chemicals
Description
A saturated fatty acid Stearic acid is used as a raw material in the production of detergents, surfactants, soaps, and cosmetics such as shampoos and shaving cream products. Its salts are used to soften polyvinyl chloride and a component of grease. It is used for the preparation of softeners in textile sizing with castor oil and dietary supplements. It acts as a hardener in candles with simple sugar or corn syrup. It is utilized for the coating of metal powders like aluminum and iron in fireworks. It finds application as a common lubricant during injection molding and pressing of ceramic powders. It acts as a key raw material for the synthesis of stearyl alcohol by reduction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
A saturated fatty acid Stearic acid is used as a raw material in the production of detergents, surfactants, soaps, and cosmetics such as shampoos and shaving cream products. Its salts are used to soften polyvinyl chloride and a component of grease. It is used for the preparation of softeners in textile sizing with castor oil and dietary supplements. It acts as a hardener in candles with simple sugar or corn syrup. It is utilized for the coating of metal powders like aluminum and iron in fireworks. It finds application as a common lubricant during injection molding and pressing of ceramic powders. It acts as a key raw material for the synthesis of stearyl alcohol by reduction.
Solubility
Soluble in ethanol, dimethyl sulfoxide, dimethyl formamide, carbon terachloride, toluene, chloroform, acetone and carbondisulfide. Insoluble in water.
Notes
Store in a cool place. Incompatible with bases, oxidizing agents and reducing agents.
Specifications
Melting Point | 69°C to 72°C |
Density | 0.941 |
Boiling Point | 232°C (15 mmHg) |
Flash Point | 196°C (384°F) |
Assay Percent Range | 98% |
Quantity | 25 g |
Beilstein | 608585 |
Solubility Information | Soluble in ethanol,dimethyl sulfoxide,dimethyl formamide,carbon terachloride,toluene,chloroform,acetone and carbondisulfide. Insoluble in water. |
Formula Weight | 284.48 |
Chemical Name or Material | Stearic Acid |
RUO – Research Use Only