missing translation for 'onlineSavingsMsg'
Learn More

Stearalkonium Chloride, TRC

Product Code. 30416859
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30416859 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30416859 Supplier Toronto Research Chemicals Supplier No. TRCS68570010G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Tilføj til indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolitik

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Stearalkonium Chloride
CAS 122-19-0
Purity Grade Notes HPLC
Molecular Formula C27 H50 N . Cl
Quantity 10 g
Synonym Benzenemethanaminium','N,N-dimethyl-N-octadecyl-','chloride (1:1), Ammonium','benzyldimethyloctadecyl-','chloride (8CI), Benzenemethanaminium','N,N-dimethyl-N-octadecyl-','chloride (9CI), Benzyldimethyloctadecylammonium chloride (6CI), 2B, 2B (onium compound), Algene SC 25, Alkaquat DMB-ST, Ammonyx 4, Ammonyx 4002, Ammonyx 490, Antistatic 1827, Arquad DM 18B90, Benzyldimethylstearylammonium chloride, Benzyloctadecyldimethylammonium chloride, Benzylstearyldimethylammonium chloride, Carsoquat SDQ 25, Carsoquat SDQ 85, Catinal OB 80E, Cation S (surfactant), Cation S 2-100, DC, DC (leveling agent), Dimethylbenzyloctadecylammonium chloride, Dimethyloctadecylbenzylammonium chloride, Dimethylstearylbenzylammonium chloride, Dodigen 1828, Incroquat S 85, Incroquat SDQ 25, Intexan SB 85, Intexsan SB 85, Larostat 905, Larostat HTS 905, Leveling agent 1827, Mackernium SDC 25, Mackernium SDC 85, Maquat SC 18, N,N-Dimethyl-N-benzyl-N-octadecylammonium chloride, N-Octadecyl-N-benzyl-N,N-dimethylammonium chloride, Nissan Cation S2-100, ODBAC 1827, Octadecylbenzyldimethylammonium chloride, Octadecyldimethylbenzylammonium chloride, Orthosan MB, Pentonium SD 75, Quaternol 1, SDQ 25, SDQ 85, Stearalkonium chloride, Stearylbenzyldimethyl ammonium chloride, Stearyldimethylbenzylammonium chloride, Stebac, Stedbac, Surfactant 1827, Swanol CA 1485, Tetranyl B 2MTH, Triton CG 400, Triton CG 500, Triton X 40, Triton X 400, Varisoft SDC, Varisoft SDC 85
InChI Formula InChI=1S/C27H50N.ClH/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-22-25-28(2,3)26-27-23-20-19-21-24-27;/h19-21,23-24H,4-18,22,25-26H2,1-3H3;1H/q+1;/p-1
SMILES [Cl-].CCCCCCCCCCCCCCCCCC[N+](C)(C)Cc1ccccc1
IUPAC Name benzyl-dimethyl-octadecylazanium;chloride
Molecular Weight (g/mol) 424.15
Formula Weight 423.3632
Percent Purity >95
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.