missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEAP3/TSAP6 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-76824CL1
This item is not returnable.
View return policy
Description
STEAP3/TSAP6 Polyclonal antibody specifically detects STEAP3/TSAP6 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| STEAP3/TSAP6 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| dudlin 2, dudlin-2, dudulin 2, dudulin-2, Dudulin-2,1010001D01Rik, EC 1.16.1.-, hpHyde, hTSAP6, metalloreductase STEAP3, pHyde, six transmembrane prostate protein 3, Six-transmembrane epithelial antigen of prostate 3, STEAP family member 3, TSAP6STMP3, tumor suppressor pHyde, Tumor suppressor-activated pathway protein 6 | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human STEAP3. The immunogen is located within the first 50 amino acids of STEAP3. Amino Acid Squence: DSDSSLAKVPDEAPK | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Endocrinology, Signal Transduction | |
| 55240 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction