missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STEAP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12818592
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12818592 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12818592 Supplier Invitrogen™ Supplier No. PA520405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is human prostate tissue lysate. PA5-20405 can be used with blocking peptide PEP-0522.

The six-transmembrane epithelial antigen of prostate 2 (STEAP2) is a member of a family of metalloreductases identified as cell-surface antigens in prostate tissue. Similar to two other members of the STEAP family (STEAP 3 and STEAP4), STEAP2 promotes both iron and copper reduction. STEAP2 expression in transfected cells also correlated with iron or copper uptake, suggesting that the STEAP family of proteins may function to stimulate iron and copper uptake. STEAP2 is widely expressed in many tissues in the plasma membrane, but is most highly expressed in prostate. At least three isoforms of STEAP2 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STEAP2
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene STEAP2
Gene Accession No. Q8BWB6, Q8NFT2
Gene Alias 4921538B17Rik; AI930049; AW045895; IPCA1; IPCA-1; Metalloreductase STEAP2; PCANAP1; prostate cancer associated protein 1; prostate cancer-associated protein 1; protein upregulated in metastatic prostate cancer; Protein up-regulated in metastatic prostate cancer; PUMPCn; six transmembrane epithelial antigen of prostate 2; six transmembrane epithelial antigen of the prostate 2; six-transmembrane epithelial antigen of prostate 2; SixTransMembrane protein of prostate 1; STAMP1; STEAP family member 2, metalloreductase; Steap2; STEAP2 metalloreductase; STMP; UNQ6507/PRO23203
Gene Symbols STEAP2
Host Species Rabbit
Immunogen A 16 amino acid peptide from near the carboxy terminus of human STEAP2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 261729, 74051
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.