missing translation for 'onlineSavingsMsg'
Learn More

STEAP2 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18408614 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408614 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408614 Supplier Novus Biologicals Supplier No. NBP176823DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

STEAP2 Polyclonal antibody specifically detects STEAP2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen STEAP2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias EC 1.16.1, EC 1.16.1.-, IPCA1, metalloreductase STEAP2, PCANAP1STMP, prostate cancer associated protein 1, Prostate cancer-associated protein 1, Protein upregulated in metastatic prostate cancer, PUMPCn, six transmembrane epithelial antigen of prostate 2, six transmembrane epithelial antigen of the prostate 2, Six-transmembrane epithelial antigen of prostate 2, SixTransMembrane protein of prostate 1, STAMP1IPCA-1
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human STEAP2. The immunogen is located within the last 50 amino acids of STEAP2. Amino Acid Squence: KLKRIKKGWEKSQFLE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 261729
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.