missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEAP2 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76823AF488
This item is not returnable.
View return policy
Description
STEAP2 Polyclonal antibody specifically detects STEAP2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| STEAP2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 261729 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 488 | |
| EC 1.16.1, EC 1.16.1.-, IPCA1, metalloreductase STEAP2, PCANAP1STMP, prostate cancer associated protein 1, Prostate cancer-associated protein 1, Protein upregulated in metastatic prostate cancer, PUMPCn, six transmembrane epithelial antigen of prostate 2, six transmembrane epithelial antigen of the prostate 2, Six-transmembrane epithelial antigen of prostate 2, SixTransMembrane protein of prostate 1, STAMP1IPCA-1 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human STEAP2. The immunogen is located within the last 50 amino acids of STEAP2. Amino Acid Squence: KLKRIKKGWEKSQFLE | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction