missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STEAP1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12808592
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12808592 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12808592 Supplier Invitrogen™ Supplier No. PA520404

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is human spleen tissue lysate. PA5-20404 can be used with blocking peptide PEP-0521.

STEAP1 is predominantly expressed in prostate tissue, and is found to be upregulated in multiple cancer cell lines. The gene product is predicted to be a six-transmembrane protein, and was shown to be a cell surface antigen significantly expressed at cell-cell junctions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STEAP1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene STEAP1
Gene Accession No. Q9CWR7, Q9UHE8
Gene Alias 2410007B19Rik; metalloreductase STEAP1; PRSS24; six transmembrane epithelial antigen of the prostate 1; six-transmembrane epithelial antigen of prostate 1; Steap; Steap1; STEAP-1; tcag7.1170
Gene Symbols STEAP1
Host Species Rabbit
Immunogen A 16 amino acid peptide from near the center of human STEAP1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26872, 297738, 70358
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.