missing translation for 'onlineSavingsMsg'
Learn More

STEAP1 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18420134 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420134 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420134 Supplier Novus Biologicals Supplier No. NBP176822G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

STEAP1 Polyclonal antibody specifically detects STEAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen STEAP1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias EC 1.16.1.-, PRSS24MGC19484, six transmembrane epithelial antigen of the prostate, six transmembrane epithelial antigen of the prostate 1, Six-transmembrane epithelial antigen of prostate 1, STEAPmetalloreductase STEAP1
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the center of human STEAP1. The immunogen is located within amino acids 130 - 180 of STEAP1. Amino Acid Squence: LHNGTKYKKFPHWLDK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 26872
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.