missing translation for 'onlineSavingsMsg'
Learn More

STEAP1 Antibody [Alexa Fluor™ 647], Novus Biologicals™

Product Code. 30713228 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30713228 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30713228 Supplier Novus Biologicals Supplier No. NBP348437AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

STEAP1 Polyclonal antibody specifically detects STEAP1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Flow Cytometry (Intracellular Staining),Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen STEAP1
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry (Intracellular Staining), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias EC 1.16.1.-, PRSS24MGC19484, six transmembrane epithelial antigen of the prostate, six transmembrane epithelial antigen of the prostate 1, Six-transmembrane epithelial antigen of prostate 1, STEAPmetalloreductase STEAP1
Host Species Rabbit
Immunogen Partial recombinant protein made to an N-terminal portion of the human STEAP protein [UniProt Q9UHE8].
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 26872
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.