missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ SteamScrubber Starter Kit

Product Code. 17789827 Shop All Labconco Corporation Products
Click to view available options
:
Starter Kit
This item is not returnable. View return policy

Product Code. 17789827

Brand: Labconco™ 4577100

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Outfit a SteamScrubber Glassware Washer to wash a variety of wide-mouth labware.

  • Use to wash a variety of wide-mouth labware
  • Includes Bulk Tube Insert (4573500), Utensil Basket with Cover (4573100), 32-Pin Insert (4401801), and 48-Pin Insert (4574200)

Specifications

Material Stainless Steel
Product Type Starter Kit
Color Silver
No. of Pins 32 or 48
Rack Type Standard
For Use With (Application) Washing Glassware
For Use With (Equipment) SteamScrubber Glassware Washer
Includes Bulk Tube Insert (4573500), Utensil Basket with Cover (4573100), 32-Pin Insert (4401801), and 48-Pin Insert (4574200)
Dimensions (L x W x H) 17.7 x 5.1 x 8.9 in.
Weight (English) Shipping 22 lb.
Weight (Metric) Shipping 9.97 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.