missing translation for 'onlineSavingsMsg'
Learn More

STCH Mouse anti-Human, Clone: 2F6, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-60243-50UL

 View more versions of this product

Product Code. 18231572

  • £240.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

STCH Monoclonal specifically detects STCH in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

STCH
Monoclonal
1 mg/mL
Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100
heat shock 70 kDa protein 13, heat shock protein 70kDa family, member 13, microsomal stress 70 protein ATPase core, Microsomal stress-70 protein ATPase core, STCHStress-70 protein chaperone microsome-associated 60 kDa protein, stress 70 protein chaperone, microsome-associated, 60kD, stress 70 protein chaperone, microsome-associated, 60kDa
Mouse
Protein A purified
RUO
6782
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1 κ
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
2F6
Unconjugated
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
HSPA13
Recombinant human HSPA13 (23-471aa) purified from E. coli
50 μL
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.