missing translation for 'onlineSavingsMsg'
Learn More

STCH Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£342.00 - £470.00

Specifications

Antigen STCH
Dilution Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:1000-1:2500
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18450231
View Documents
Novus Biologicals
NBP1-80715-25ul
25ul
£342.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18258395
View Documents
Novus Biologicals
NBP1-80715
0.1 mL
£470.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

STCH Polyclonal specifically detects STCH in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

STCH
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
P48723
6782
This antibody was developed against Recombinant Protein corresponding to amino acids:AYGLHKADVFHVLVIDLGGGTLDVSLLNKQGGMFLTRAMSGNNKLGGQDFNQRLLQYLYKQIYQTYGFVPSRKEEIHRLRQAVEMVKLNLTLHQSAQLSVLLTVEEQDRKEPHSSDTELPKDKLSSADDH
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:1000-1:2500
Polyclonal
Rabbit
Signal Transduction
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
heat shock 70 kDa protein 13, heat shock protein 70kDa family, member 13, microsomal stress 70 protein ATPase core, Microsomal stress-70 protein ATPase core, STCHStress-70 protein chaperone microsome-associated 60 kDa protein, stress 70 protein chaperone, microsome-associated, 60kD, stress 70 protein chaperone, microsome-associated, 60kDa
HSPA13
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.