missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583770
This item is not returnable.
View return policy
Description
Immunogen sequence: SAIQKPPTAP PERQPQVDRT KLSRAHHGEA GHHLPEPSSR ETGRGAKGER GSKSHPNAHA RGRVGGLGAQ GPSGSSEWED EQSEYSDIRR Highest antigen sequence indentity to the following orthologs: Rat - 68%, Mouse - 72%.
Has an anti-hypocalcemic action on calcium and phosphate homeostasis.
Specifications
| STC2 | |
| Polyclonal | |
| Unconjugated | |
| STC2 | |
| AW125853; mustc2; stanniocalcin 2; stanniocalcin-2; Stanniocalcin-related protein; STC2; STC-2; Stc2l; STC-related protein; STCRP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8614 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O76061 | |
| STC2 | |
| Recombinant protein corresponding to Human STC2. Recombinant protein control fragment (Product #RP-100450). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction