missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STC2 Monoclonal Antibody (348217)
Rat Monoclonal Antibody
Brand: Invitrogen™ MA561210
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Has an anti-hypocalcemic action on calcium and phosphate homeostasis.
Specifications
| STC2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O76061 | |
| STC2 | |
| Mouse myeloma cell line NS0-derived recombinant human Stanniocalcin 2, Thr25-Arg302. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 348217 | |
| Unconjugated | |
| STC2 | |
| AW125853; mustc2; stanniocalcin 2; stanniocalcin-2; Stanniocalcin-related protein; STC2; STC-2; Stc2l; STC-related protein; STCRP | |
| Rat | |
| Protein A/G | |
| RUO | |
| 8614 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction