missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103085
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Stanniocalcin 1.
This gene encodes a secreted, homodimeric glycoprotein that is expressed in a wide variety of tissues and may have autocrine or paracrine functions. The gene contains a 5' UTR rich in CAG trinucleotide repeats. The encoded protein contains 11 conserved cysteine residues and is phosphorylated by protein kinase C exclusively on its serine residues. The protein may play a role in the regulation of renal and intestinal calcium and phosphate transport, cell metabolism, or cellular calcium/phosphate homeostasis. Overexpression of human stanniocalcin 1 in mice produces high serum phosphate levels, dwarfism, and increased metabolic rate. This gene has altered expression in hepatocellular, ovarian, and breast cancers.
Specifications
| STC1 | |
| Polyclonal | |
| Unconjugated | |
| STC1 | |
| stanniocalcin; stanniocalcin 1; stanniocalcin-1; STC; STC1; STC-1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6781 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P52823 | |
| STC1 | |
| A synthesized peptide derived from human STC1(Accession P52823), corresponding to amino acid residues K185-S235. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction