missing translation for 'onlineSavingsMsg'
Learn More

STB5L Polyclonal Antibody, Invitrogen™

Product Code. 11541302
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11541302 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11541302 Supplier Invitrogen Supplier No. OSS00049W

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reconstitute with 100 μL of distilled water.

TRUSTED_SUSTAINABILITY

Specifications

Antigen STB5L
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation whole serum with no preservative
Gene Stxbp5l
Gene Accession No. Q5DQR4
Gene Alias Tomosyn-2, Lethal(2) giant larvae protein homolog 4, Stxbp5l, Llgl4, LLGL4, STB5L, MGC169507, 9430094L02, A830015P08Rik
Gene Symbols STXBP5L
Host Species Rabbit
Immunogen A synthetic peptide Rabbit antibody to c-terminal region of mouse Syntaxin-binding protein 5-like
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 207227, 288080
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.