missing translation for 'onlineSavingsMsg'
Learn More

Stavudine, TRC

Product Code. 30423514
Change view
Click to view available options
Quantity:
10 mg
25 mg
1 g
Product Code. Quantity
30423514 1 g
30412449 25 mg
30415525 10 mg
3 options
This item is not returnable. View return policy

Product Code. 30423514

Brand: Toronto Research Chemicals TRCS6852501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3056-17-5
Formula Weight 224.0797
InChI Formula InChI=1S/C10H12N2O4/c1-6-4-12(10(15)11-9(6)14)8-3-2-7(5-13)16-8/h2-4,7-8,13H,5H2,1H3,(H,11,14,15)/t7-,8+/m0/s1
IUPAC Name 1-[(2R,5S)-5-(hydroxymethyl)-2,5-dihydrofuran-2-yl]-5-methylpyrimidine-2,4-dione
Molecular Formula C10 H12 N2 O4
Molecular Weight (g/mol) 224.21
SMILES CC1=CN([C@@H]2O[C@H](CO)C=C2)C(=O)NC1=O
Synonym Stavudine, 1-[(2R,5S)-5-(Hydroxymethyl)-2,5-dihydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Zidovudine Imp. A (EP), 1-(2,3-Dideoxy-β-D-glycero-pent-2-enofuranosyl)thymine, 3'-Deoxy-2'-thymidinene, 2',3'-Didehydro-3'-deoxythymidine, 3'-Deoxy-2',3'-didehydrothymidine, Avostav, BMY 27857, D 4T (nucleoside), NSC 163661, Sanilvudine, Staduvine, Stag, Stavir, Virostav, Zerit, d4T
Chemical Name or Material Stavudine

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.