missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Staurosporine, 98%

Product Code. 10743783 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mg
250 μg
Unit Size:
1mg
250µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10743783 250 μg 250µg
10269653 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10743783 Supplier Thermo Scientific Chemicals Supplier No. 328532500
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Staurosporine (staurosporin, CAS # 62996-74-1) is a bacterial alkyloid isolated from Streptomyces staurosporeus.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Staurosporine is an indolocarbazole alkaloid capable of permeating cell membranes
  • Staurosporine has anti-cancer properties that can induce apoptosis
  • Staurosporine is a broad inhibitor of protein kinases, preventing growth and division cycles

Application

  • Staurosporine is used for research on acute myeloid leukemia
  • It is suitable for research applications including cell culture, in vivo studies, and biochemistry
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62996-74-1
Molecular Formula C28H26N4O3
Molecular Weight (g/mol) 466.5
MDL Number MFCD00077402
InChI Key HKSZLNNOFSGOKW-ZYSRIHRCSA-N
Synonym staurosporine, kinome_3629
PubChem CID 49831000
SMILES CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC

Specifications

Melting Point 270.0°C
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 μg
Merck Index 15,8928
Solubility Information Soluble in DMSO, ethyl acetate or dimethyl formamide. Insoluble in water.
Formula Weight 466.5
Percent Purity 98%
Chemical Name or Material Staurosporine
compliance-icons
Product Identifier
  • Staurosporine
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.