missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Staurosporine

Product Code. 30001226
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001226 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001226 Supplier R&D Systems™ Supplier No. 1285/100U
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Non-selective protein kinase inhibitor

Staurosporine is a broad spectrum protein kinase inhibitor. Enzymes inhibited include protein kinase C, p60v-src tyrosine protein kinase, protein kinase A, and CaM kinase II (IC50 values are 3nM, 6 nM, 7 nM and 20 nM, respectively). Staurosporine reduces nuclear myosin heavy chain 9 phosphorylation which inhibits gastric cancer cell progression in transgenic mouse models. Staurosporine inhibits cell viability and promotes apoptosis in oral and pancreatic cancer cells. Staurosporine also enhances efficiency of lentiviral transduction of human hematopoietic stem and progenitor cells by 2-fold, induces dopaminergic axonal outgrowth in vitro and triggers mitophagy. Staurosporine has high affinity (Kd = 100 nM) for the yjdF aptamer. The riboswitch function of yjdF motif RNAs is activated by Staurosporine and leads to robust reporter gene expressions in B. subtilis.

Staurosporine synthesized to Ancillary Material Grade is also available.

TRUSTED_SUSTAINABILITY

Specifications

CAS 62996-74-1
Quantity 100 μg
Product Line Tocris
Molecular Formula C28H26N4O3
Formula Weight 466.54
Purity >98%
Molecular Weight (g/mol) 466.541
Chemical Name or Material Staurosporine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.