missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Staufen Rabbit anti-Human, Mouse, Rat, Clone: 8J5A6, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Staufen Monoclonal antibody specifically detects Staufen in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Staufen |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 8J5A6 |
| Conjugate | Unconjugated |
| Formulation | PBS, 0.05% BSA, 50% glycerol, pH7.3 |
| Gene Alias | double-stranded RNA-binding protein Staufen homolog 1, FLJ25010, RNA binding protein (Drosophila), RNA-binding protein), staufen, RNA binding protein, homolog 1 (Drosophila) |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 478-577 of human Staufen (O95793). |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?