missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
STAU2 Monoclonal antibody specifically detects STAU2 in Human samples. It is validated for Western Blot, ELISA, ELISA
Specifications
Specifications
| Antigen | STAU2 |
| Applications | Western Blot, ELISA, Sandwich ELISA |
| Classification | Monoclonal |
| Clone | 3E9 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_055208 |
| Gene Alias | DKFZp781K0371,39K2, double-stranded RNA-binding protein Staufen homolog 2,39K3, MGC119606, staufen (Drosophila, RNA-binding protein) homolog 2, staufen homolog 2, staufen, RNA binding protein, homolog 2 (Drosophila) |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?