missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAU1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14225-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the DEAD box protein family. The protein is an RNA helicase found in P-bodies and stress granules, and functions in translation suppression and mRNA degradation. It is required for microRNA-induced gene silencing.Specifications
| STAU1 | |
| Polyclonal | |
| Unconjugated | |
| STAU1 | |
| STAU, STAU1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20853, 6780, 84496 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95793, Q9Z108 | |
| Stau1 | |
| STAU1 Fusion Protein Ag5464 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction