Description
- Immediately recognize when determinations are finished and react to warnings accordingly
- Green - The system is ready for operation
- Yellow - The system is operating
- Red - When there is an error
Specifications
Specifications
| Includes | Status Light |