missing translation for 'onlineSavingsMsg'
Learn More

Stattic, MedChemExpress

Product Code. 16452027
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 16452027

missing translation for 'mfr': Medchem Express HY13818

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Stattic is a potent STAT3 inhibitor and inhibits STAT3 phosphorylation (at Y705 and S727). Stattic inhibits the binding of a high affinity phosphopeptide for the SH2 domain of STAT3. Stattic ameliorates the renal dysfunction in Alport syndrome (AS) mice.

Stattic is a potent STAT3 inhibitor and inhibits STAT3 phosphorylation (at Y705 and S727). Stattic inhibits the binding of a high affinity phosphopeptide for the SH2 domain of STAT3. Stattic ameliorates the renal dysfunction in Alport syndrome (AS) mice.

Specifications

Chemical Name or Material Stattic
CAS 19983-44-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C8H5NO4S
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (236.75 mM; Need ultrasonic)
SMILES O=[N+](C1=CC=C(C=CS2(=O)=O)C2=C1)[O-]
Molecular Weight (g/mol) 211.19
Formula Weight 211.19
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.