missing translation for 'onlineSavingsMsg'
Learn More

Stattic, MedChemExpress

Product Code. 16483386
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483386 25 mg 25mg
16452027 50 mg 50mg
16445878 10 mg 10mg
16486336 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16483386 Supplier Medchem Express Supplier No. HY13818
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Stattic is a potent STAT3 inhibitor and inhibits STAT3 phosphorylation (at Y705 and S727). Stattic inhibits the binding of a high affinity phosphopeptide for the SH2 domain of STAT3. Stattic ameliorates the renal dysfunction in Alport syndrome (AS) mice.

Stattic is a potent STAT3 inhibitor and inhibits STAT3 phosphorylation (at Y705 and S727). Stattic inhibits the binding of a high affinity phosphopeptide for the SH2 domain of STAT3. Stattic ameliorates the renal dysfunction in Alport syndrome (AS) mice.

Specifications

Chemical Name or Material Stattic
CAS 19983-44-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C8H5NO4S
Quantity 25 mg
Solubility Information DMSO : 50 mg/mL (236.75 mM; Need ultrasonic)
SMILES O=[N+](C1=CC=C(C=CS2(=O)=O)C2=C1)[O-]
Molecular Weight (g/mol) 211.19
Formula Weight 211.19
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.