Learn More
Abeba™ Static Control 31392 Shoes
Product Code. 15618907
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
47
Unit Size:
1 pair
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15618907 45 1 pair
15628897 36 1 pair
15638897 37 1 pair
15648897 38 1 pair
15658897 39 1 pair
15668897 40 1 pair
15678897 41 1 pair
15688897 42 1 pair
15698897 43 1 pair
15608907 44 1 pair
15638907 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15618907 Supplier Abeba™ Supplier No. 31392/45
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
ESD dissipative and Sanitized™ treated.
  • CE EN ISO 20345:2011 S2 SRA
  • Microfiber
  • White
  • Composite toe cap
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Replaceable acc Wave insole (article no. 3553)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with elastic band
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S2 SRA
Color White
Material Microfiber
Size 45
Sole Features Slip-resistant
Sole Material Rubber
Style 31392
Toe Material Composite