missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stathmin-2/STMN2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-1403
This item is not returnable.
View return policy
Description
Stathmin-2/STMN2 Polyclonal specifically detects Stathmin-2/STMN2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| Stathmin-2/STMN2 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| FLJ34868, FLJ50995, neuron-specific growth-associated protein, Protein SCG10, SCG10, SCG10neuronal growth-associated protein (silencer element), SCGN10stathmin-2, stathmin-like 2, superior cervical ganglia, neural specific 10, Superior cervical ganglion-10 protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 11075 | |
| Mouse | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:128000 | |
| NP_008960. | |
| STMN2 | |
| Peptide with sequence AKTAMAYKEK-C corresponding to N-Terminus according to NP_008960.2. | |
| 0.1 mg | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse, Rat and Dog. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction