missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stathmin 1 Antibody (STMN1/8023R) [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20919JF635
This item is not returnable.
View return policy
Description
Stathmin 1 Monoclonal antibody specifically detects Stathmin 1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Stathmin 1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| C1orf215, chromosome 1 open reading frame 215, Lag, Leukemia-associated phosphoprotein p18, Metablastin, MGC138869, Oncoprotein 18, Op18, OP18MGC138870, phosphoprotein 19, Phosphoprotein p19, pp17, PP19, PR22, prosolin, Protein Pr22, SMNLAP18FLJ32206, stathmin, stathmin 1, stathmin 1/oncoprotein 18, transmembrane protein C1orf215 | |
| Recombinant fragment of human Stathmin 1 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cognition and Behavior, Immunology, Microglia Markers, Neuroscience, Phospho Specific, Signal Transduction | |
| 3925 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| STMN1/8023R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction