missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Stathmin 1 Antibody (rSTMN1/9255) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26959PE
This item is not returnable.
View return policy
Description
Stathmin 1 Monoclonal antibody specifically detects Stathmin 1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Stathmin 1 | |
| Monoclonal | |
| PE | |
| C1orf215, chromosome 1 open reading frame 215, Lag, Leukemia-associated phosphoprotein p18, Metablastin, MGC138869, Oncoprotein 18, Op18, OP18MGC138870, phosphoprotein 19, Phosphoprotein p19, pp17, PP19, PR22, prosolin, Protein Pr22, SMNLAP18FLJ32206, stathmin, stathmin 1, stathmin 1/oncoprotein 18, transmembrane protein C1orf215 | |
| Recombinant fragment (around aa1-148) of human Stathmin 1 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cognition and Behavior, Immunology, Microglia Markers, Neuroscience, Phospho Specific, Signal Transduction | |
| 3925 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| rSTMN1/9255 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction