missing translation for 'onlineSavingsMsg'
Learn More

Stathmin 1 Antibody, DyLight 550, Novus Biologicals™

Product Code. 18375933 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18375933 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18375933 Supplier Novus Biologicals Supplier No. NBP299433R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Stathmin 1 Polyclonal specifically detects Stathmin 1 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Stathmin 1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias C1orf215, chromosome 1 open reading frame 215, Lag, Leukemia-associated phosphoprotein p18, Metablastin, MGC138869, Oncoprotein 18, Op18, OP18MGC138870, phosphoprotein 19, Phosphoprotein p19, pp17, PP19, PR22, prosolin, Protein Pr22, SMNLAP18FLJ32206, stathmin, stathmin 1, stathmin 1/oncoprotein 18, transmembrane protein C1orf215
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of mouse Stathmin 1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cognition and Behavior, Immunology, Microglia Markers, Neuroscience, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3925
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.