missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT5b Rabbit anti-Human, Mouse, Rat, Clone: 6P5T0, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £425.00
Specifications
| Antigen | STAT5b |
|---|---|
| Clone | 6P5T0 |
| Applications | Western Blot, Immunoprecipitation, KnockDown |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18336435
|
Novus Biologicals
NBP3-15778-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18345154
|
Novus Biologicals
NBP3-15778-100UL |
100 μg |
£425.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
STAT5b Monoclonal antibody specifically detects STAT5b in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation, KnockDownSpecifications
| STAT5b | |
| Western Blot, Immunoprecipitation, KnockDown | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 6777 | |
| IgG | |
| Affinity purified |
| 6P5T0 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 600-787 of human STAT5b (P51692). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title