missing translation for 'onlineSavingsMsg'
Learn More

STAT5b Antibody (STAT5B/2657) - Azide and BSA Free, Novus Biologicals™

Product Code. 18342672 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342672 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18342672 Supplier Novus Biologicals Supplier No. NBP308628

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

STAT5b Monoclonal specifically detects STAT5b in Human samples. It is validated for ELISA, Protein Array.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAT5b
Applications ELISA, Peptide Array
Classification Monoclonal
Clone STAT5B/2657
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA : 1-5 ug/mL, Protein Array
Formulation 10mM PBS
Gene Alias signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B
Host Species Mouse
Immunogen Recombinant full-length human STAT5b protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6777
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.