missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT5b Antibody (389215) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1584JF669
This item is not returnable.
View return policy
Description
STAT5b Monoclonal antibody specifically detects STAT5b in Human samples. It is validated for Immunoprecipitation,Western Blot,Simple Western,Flow Cytometry (Intracellular Staining),CyTOF,Gene Knock-Out
Specifications
| STAT5b | |
| Monoclonal | |
| Janelia Fluor 669 | |
| signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B | |
| Human STAT5b synthetic peptide, MDSQWIPHAQS, Accession # NP_036580 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Immunoprecipitation, Western Blot, Simple Western, Flow Cytometry (Intracellular Staining), CyTOF, Gene Knock-Out | |
| 389215 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Phospho Specific, Signal Transduction | |
| 6777 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction