missing translation for 'onlineSavingsMsg'
Learn More

STAT5b Antibody (389215) [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30169167 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30169167 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30169167 Supplier Novus Biologicals Supplier No. FAB1584JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

STAT5b Monoclonal antibody specifically detects STAT5b in Human samples. It is validated for Immunoprecipitation,Western Blot,Simple Western,Flow Cytometry (Intracellular Staining),CyTOF,Gene Knock-Out
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAT5b
Applications Immunoprecipitation, Western Blot, Simple Western, Flow Cytometry (Intracellular Staining), CyTOF, Gene Knock-Out
Classification Monoclonal
Clone 389215
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B
Host Species Mouse
Immunogen Human STAT5b synthetic peptide, MDSQWIPHAQS, Accession # NP_036580
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6777
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.