missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT5A, p Tyr694 Antibody (5F6.F1), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-77817AF647
This item is not returnable.
View return policy
Description
STAT5A Monoclonal specifically detects STAT5A in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| STAT5a (p Tyr694) | |
| Monoclonal | |
| Alexa Fluor 647 | |
| MGF, signal transducer and activator of transcription 5A, STAT5 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody is specific for mouse Stat5a protein phosphorylated at Y694. A BLAST analysis was used to suggest cross-reactivity with Stat5a from human, mouse and rat based on 100% homology with the immunizing sequence. Cross-reactivity with Stat5a from other sources has not been determined. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 5F6.F1 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| STAT5A | |
| STAT5AA [p Tyr694] antibody (5F6.F1) was produced by repeated immunizations with a synthetic peptide corresponding to residues surrounding Y694 of mouse STAT5Aa protein. (Uniprot: P42230) | |
| 0.1 mL | |
| Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 6776 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction