missing translation for 'onlineSavingsMsg'
Learn More

STAT5A, p Tyr694 Antibody (5F6.F1), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18403577 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403577 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403577 Supplier Novus Biologicals Supplier No. NBP177817AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

STAT5A Monoclonal antibody specifically detects STAT5A in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAT5A
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5F6.F1
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias MGF, signal transducer and activator of transcription 5A, STAT5
Host Species Mouse
Immunogen STAT5AA [p Tyr694] antibody (5F6.F1) was produced by repeated immunizations with a synthetic peptide corresponding to residues surrounding Y694 of mouse STAT5Aa protein. (Uniprot: P42230)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6776
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.