missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT5a/b, p Tyr694, p Tyr699 Antibody (1247C), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB41901MFV450
This item is not returnable.
View return policy
Description
STAT5a/b Monoclonal antibody specifically detects STAT5a/b in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| STAT5a/b | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 6776 | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, CyTOF | |
| 1247C | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| MGF, STAT5, STAT5A signal transducer and activator of transcription 5A | |
| Phosphopeptide containing human STAT5b Y699 site (amino acid sequence of this peptide is identical to a corresponding region of human STAT5a containing Y694), Accession # P51692 | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction