missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT5A Antibody (6D4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37441
This item is not returnable.
View return policy
Description
STAT5A Monoclonal specifically detects STAT5A in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| STAT5a | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready | |
| MGF, signal transducer and activator of transcription 5A, STAT5 | |
| Mouse | |
| 90.6 kDa | |
| 0.1 mL | |
| Cancer, Signal Transduction, Transcription Factors and Regulators | |
| 6776 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 6D4 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| STAT5A | |
| Purified recombinant fragment of human STAT5A (AA: 583-794 ) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction