missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STAT5 alpha (Total/Phospho) [pY694/pY699] Human InstantOne™ ELISA Kit

InstantOne ELISA

Brand:  Invitrogen™ 85-86113-11

Additional Details : Weight : 0.30000kg

Product Code. 15974194

  • £534.00 / Pack of 96
Estimated Shipment: 30-05-2024
to see stock.
Add to basket

Description

Description

The InstantOne™ ELISA is specifically engineered for accurate measurement of total and phosphorylated human STAT5A/B (Tyr694/Tyr699) in cell lysates. Detection of mouse and rat STAT5A/B is expected. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.

STAT5 alpha is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. STAT5 alpha is activated by, and mediates the responses of many cell ligands, such as IL2, IL3, IL7 GM-CSF, erythropoietin, thrombopoietin, and different growth hormones. Activation of STAT5 alpha in myeloma and lymphoma associated with a TEL/JAK2 gene fusion is independent of cell stimulus and has been shown to be essential for the tumorigenesis. The mouse counterpart of STAT5 alpha is found to induce the expression of BCL2L1/BCL-X(L), which suggests the antiapoptotic function of this protein in cells. STAT5 alpha, along with STAT5 beta, were identified in mouse. The amino acid sequences of STAT5 alpha and STAT5 beta show 96% sequence similarity, and both proteins are co expressed in most tissues in virgin and lactating mice. However, differential accumulation of STAT5 alpha and STAT5 beta mRNA has been reported for both muscle and mammary tissue. STAT5 alpha is critically involved in a variety of physiological functions, including reproduction, lactation, immune function and somatic growth.
Specifications

Specifications

P42229
ELISA Kit
Human
6776
Capture Antibody, Detection Antibody Reagent, InstantOne™ Assay Plate, Cell Lysis Buffer, Enhancer Solution, Wash Buffer (10X), Substrate Reagent (TMB), Stop Solution, Positive Control Cell Lysate (lyophilized), Plate seal
96 Tests
Cell Lysate,20 μL
Human
1 hr. 10 min.
Biotin
Cell Lysates
Colorimetric Microplate Reader
MGF,STAT5
HRP
RUO
2°C to 8°C
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers